COM497527160 Combi-Blocks, Inc.

2-Dimethylamino-benzoic acid ethyl ester

  • MDL number MFCD00009110
  • Cas Number : 55426-74-9
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8943
Availability details for
Quantity
  • Availability
  • Add to Cart