COM497527188 Combi-Blocks, Inc.

3-(Bromomethyl)quinoxalin-2(1h)-one

  • MDL number MFCD02660613
  • Cas Number : 62235-61-4
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-9633
Availability details for
Quantity
  • Availability
  • Add to Cart