COM497527203 Combi-Blocks, Inc.

3-Amino-2-phenyl-pyridine, HCl

  • MDL number MFCD03839820
  • Cas Number : 219121-62-7
  • Molecular Weight : 206.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0025
Availability details for
Quantity
  • Availability
  • Add to Cart