COM497527294 Combi-Blocks, Inc.

4-Aminomethyltetrahydropyran-4-carboxylic acid

  • MDL number MFCD09800381
  • Cas Number : 948015-51-8
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2808
Availability details for
Quantity
  • Availability
  • Add to Cart