COM497527385 Combi-Blocks, Inc.

6-(Propan-2-yl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD11616599
  • Cas Number : 570416-36-3
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5289
Availability details for
Quantity
  • Availability
  • Add to Cart