COM497527435 Combi-Blocks, Inc.

Alpha-chloro-9-anthraldoxime

  • MDL number MFCD02179418
  • Cas Number : 113003-49-9
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7014
Availability details for
Quantity
  • Availability
  • Add to Cart