COM497527449 Combi-Blocks, Inc.

Cyclooctanecarbaldehyde

  • MDL number MFCD00001747
  • Cas Number : 6688-11-5
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7504

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7504
Availability details for
Quantity
  • Availability
  • Add to Cart