COM514239137 Combi-Blocks, Inc.

2-Benzyloxy-3-bromo-5-fluorobenzonitrile

  • MDL number MFCD30182383
  • Cas Number : 1951439-43-2
  • Molecular Weight : 306.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5920
Availability details for
Quantity
  • Availability
  • Add to Cart