COM514239210 Combi-Blocks, Inc.

4-(N-methylacetamido)benzoic acid

  • MDL number MFCD20542852
  • Cas Number : 26961-99-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1537
Availability details for
Quantity
  • Availability
  • Add to Cart