COM514239353 Combi-Blocks, Inc.

1H-Benzo[d]imidazole-6-carbonitrile

  • MDL number MFCD18807947
  • Cas Number : 6287-83-8
  • Molecular Weight : 143.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4130
Availability details for
Quantity
  • Availability
  • Add to Cart