COM514239460 Combi-Blocks, Inc.

2-(4-Bromobenzyl)thiazole-4-acetic acid

  • MDL number MFCD10568322
  • Cas Number : 1053656-92-0
  • Molecular Weight : 312.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3871
Availability details for
Quantity
  • Availability
  • Add to Cart