COM514239492 Combi-Blocks, Inc.

Ethyl 3-bromo-2-oxobutanoate

  • MDL number MFCD19440735
  • Cas Number : 57332-84-0
  • Molecular Weight : 209.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1255
Availability details for
Quantity
  • Availability
  • Add to Cart