COM514239530 Combi-Blocks, Inc.

3-Nitro-4h-1,2,4-triazole

  • MDL number MFCD00276728
  • Cas Number : 24807-55-4
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9059
Availability details for
Quantity
  • Availability
  • Add to Cart