COM514239532 Combi-Blocks, Inc.

4-Iodo-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD01459883
  • Cas Number : 6647-93-4
  • Molecular Weight : 238.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9668
Availability details for
Quantity
  • Availability
  • Add to Cart