COM514239733 Combi-Blocks, Inc.

4-(Isopropylthio)-1h-imidazole

  • MDL number MFCD20617064
  • Cas Number : 1215198-00-7
  • Molecular Weight : 142.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-2149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-2149
Availability details for
Quantity
  • Availability
  • Add to Cart