COM514239736 Combi-Blocks, Inc.

2,5-Difluoro-4-formylbenzonitrile

  • MDL number MFCD16876871
  • Cas Number : 433940-02-4
  • Molecular Weight : 167.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4189
Availability details for
Quantity
  • Availability
  • Add to Cart