COM514239797 Combi-Blocks, Inc.

4-Chloro-2-fluoro-5-methoxy benzaldehyde

  • MDL number MFCD05863886
  • Cas Number : 177034-24-1
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3001
Availability details for
Quantity
  • Availability
  • Add to Cart