COM514239848 Combi-Blocks, Inc.

Mesobiliverdin

  • MDL number MFCD22684685
  • Cas Number : 493-88-9
  • Molecular Weight : 586.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WA-4763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WA-4763
Availability details for
Quantity
  • Availability
  • Add to Cart