COM514239859 Combi-Blocks, Inc.

Deuteroporphyrin ix 2,4-disulfonic acid dimethyl ester disodium salt

  • MDL number MFCD00216777
  • Molecular Weight : 742.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WA-6015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WA-6015
Availability details for
Quantity
  • Availability
  • Add to Cart