COM517297725 Combi-Blocks, Inc.

7-Methoxy-3H-1,2,3-benzotriazin-4-one

  • MDL number MFCD30474679
  • Cas Number : 41994-97-2
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1536

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1536
Availability details for
Quantity
  • Availability
  • Add to Cart