COM517297756 Combi-Blocks, Inc.

2-(Benzyloxy)-3-bromobenzonitrile

  • MDL number MFCD28128126
  • Cas Number : 862992-93-6
  • Molecular Weight : 288.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5919
Availability details for
Quantity
  • Availability
  • Add to Cart