COM517297814 Combi-Blocks, Inc.

2-Ethylpyrazole-3-carbaldehyde

  • MDL number MFCD03130036
  • Cas Number : 902837-62-1
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2310
Availability details for
Quantity
  • Availability
  • Add to Cart