COM517297833 Combi-Blocks, Inc.

4-{[(tert-Butoxy)carbonyl]amino}phenoxyacetic acid

  • MDL number MFCD14529527
  • Cas Number : 188751-52-2
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1552
Availability details for
Quantity
  • Availability
  • Add to Cart