COM517297851 Combi-Blocks, Inc.

1-Chloro-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzene

  • MDL number MFCD08276329
  • Cas Number : 346656-42-6
  • Molecular Weight : 224.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NE-2259

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NE-2259
Availability details for
Quantity
  • Availability
  • Add to Cart