COM517297857 Combi-Blocks, Inc.

1-(Pentafluoropropionyl)imidazole

  • MDL number MFCD00014502
  • Cas Number : 71735-32-5
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2334
Availability details for
Quantity
  • Availability
  • Add to Cart