COM517297878 Combi-Blocks, Inc.

Di-phthaloyl-cystamine

  • MDL number MFCD30527657
  • Cas Number : 109653-49-8
  • Molecular Weight : 412.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4249
Availability details for
Quantity
  • Availability
  • Add to Cart