COM517297949 Combi-Blocks, Inc.

3,4-Dimethylphenyl isocyanate

  • MDL number MFCD00013867
  • Cas Number : 51163-27-0
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5989

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5989
Availability details for
Quantity
  • Availability
  • Add to Cart