COM517297967 Combi-Blocks, Inc.

2-(4-Fluorophenyl)-2-methylmorpholine

  • MDL number MFCD08061112
  • Cas Number : 109461-46-3
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1917
Availability details for
Quantity
  • Availability
  • Add to Cart