COM517298061 Combi-Blocks, Inc.

2,6-Difluoropyridine-3,5-diol

  • MDL number MFCD30182431
  • Cas Number : 2010955-41-4
  • Molecular Weight : 147.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2265
Availability details for
Quantity
  • Availability
  • Add to Cart