COM633089443 Combi-Blocks, Inc.

2-(4-Benzyloxyphenyl)-4-hydroxypyridine

  • MDL number MFCD18323816
  • Cas Number : 1261975-11-4
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4192
Availability details for
Quantity
  • Availability
  • Add to Cart