COM633089490 Combi-Blocks, Inc.

4-(2-Methylphenyl)nicotinic acid

  • MDL number MFCD18207544
  • Cas Number : 207850-76-8
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3618
Availability details for
Quantity
  • Availability
  • Add to Cart