COM633144246 Combi-Blocks, Inc.

Trans-1,4-dimethylcyclohexane

  • MDL number MFCD00064955
  • Cas Number : 2207-04-7
  • Molecular Weight : 112.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8000
Availability details for
Quantity
  • Availability
  • Add to Cart