COM633144378 Combi-Blocks, Inc.

3-(3,4-Dimethoxyphenyl)-2-hydroxypyridine

  • MDL number MFCD18312281
  • Cas Number : 1261938-85-5
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0578
Availability details for
Quantity
  • Availability
  • Add to Cart