COM633279427 Combi-Blocks, Inc.

3-Methyl-2-pentanol

  • MDL number MFCD00004528
  • Cas Number : 565-60-6
  • Molecular Weight : 102.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5616
Availability details for
Quantity
  • Availability
  • Add to Cart