COM633346135 Combi-Blocks, Inc.

Methyldimethoxysilane

  • MDL number MFCD00048019
  • Cas Number : 16881-77-9
  • Molecular Weight : 106.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7094
Availability details for
Quantity
  • Availability
  • Add to Cart