COM633346710 Combi-Blocks, Inc.

2-Amino-4-chloro-3-cyano-5-formylthiophene

  • MDL number MFCD03788265
  • Cas Number : 104366-23-6
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2245
Availability details for
Quantity
  • Availability
  • Add to Cart