COM633413381 Combi-Blocks, Inc.

4-(2-Fluoro-3-trifluoromethylphenyl)-2-methoxyphenol

  • MDL number MFCD18315332
  • Cas Number : 1262001-60-4
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4400
Availability details for
Quantity
  • Availability
  • Add to Cart