COM633414150 Combi-Blocks, Inc.

2-(4-Fluorophenyl)-2-phenylethylamine, HCl

  • MDL number MFCD02089463
  • Cas Number : 79314-49-1
  • Molecular Weight : 251.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3355

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3355
Availability details for
Quantity
  • Availability
  • Add to Cart