COM633414171 Combi-Blocks, Inc.

2-(2-Carboxyvinyl)phenylboronic acid

  • MDL number MFCD02093067
  • Cas Number : 374105-86-9
  • Molecular Weight : 191.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2174
Availability details for
Quantity
  • Availability
  • Add to Cart