COM633426286 Combi-Blocks, Inc.

5-Fluoro-2-phenylpyridine

  • MDL number MFCD11044346
  • Cas Number : 512171-81-2
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4260
Availability details for
Quantity
  • Availability
  • Add to Cart