COM633426394 Combi-Blocks, Inc.

2-Fluoro-4-(2-trifluoromethylphenyl)benzoic acid

  • MDL number MFCD18321733
  • Cas Number : 505083-00-1
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2002
Availability details for
Quantity
  • Availability
  • Add to Cart