COM633548955 Combi-Blocks, Inc.

2-(2-Chloro-5-methoxyphenyl)benzoic acid

  • MDL number MFCD18312671
  • Cas Number : 1261974-52-0
  • Molecular Weight : 262.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1273

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1273
Availability details for
Quantity
  • Availability
  • Add to Cart