COM633549191 Combi-Blocks, Inc.

2-Chloroheptane

  • MDL number MFCD00039353
  • Cas Number : 1001-89-4
  • Molecular Weight : 134.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5860
Availability details for
Quantity
  • Availability
  • Add to Cart