COM633549519 Combi-Blocks, Inc.

(3R)-(+)-3-Acetamidopyrrolidine

  • MDL number MFCD00082729
  • Cas Number : 131900-62-4
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8644
Availability details for
Quantity
  • Availability
  • Add to Cart