COM633605734 Combi-Blocks, Inc.

4-(2-Methoxyphenyl)-3-methylbenzoic acid

  • MDL number MFCD18319475
  • Cas Number : 473264-05-0
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9510
Availability details for
Quantity
  • Availability
  • Add to Cart