COM638461694 Combi-Blocks, Inc.

2,6-Difluoro-3-nitrophenol

  • MDL number MFCD28790772
  • Cas Number : 1393179-72-0
  • Molecular Weight : 175.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6189
Availability details for
Quantity
  • Availability
  • Add to Cart