COM638461699 Combi-Blocks, Inc.

N-(4-Amino-2-chloro-5-hydroxyphenyl)acetamide

  • MDL number MFCD24608066
  • Cas Number : 144680-96-6
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6204
Availability details for
Quantity
  • Availability
  • Add to Cart