COM638461711 Combi-Blocks, Inc.

(2S)-2-[(Benzyloxy)methyl]-2,3-dihydro-1,4-benzodioxin-6-amine

  • MDL number MFCD30527614
  • Cas Number : 2055114-60-6
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6228
Availability details for
Quantity
  • Availability
  • Add to Cart