COM638462110 Combi-Blocks, Inc.

(3,4-Difluoro-phenyl)-piperidin-4-yl-methanone, HCl

  • MDL number MFCD08544120
  • Cas Number : 140235-26-3
  • Molecular Weight : 261.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8599
Availability details for
Quantity
  • Availability
  • Add to Cart