COM638462132 Combi-Blocks, Inc.

2-Amino-4-bromo-3-hydroxypyridine hbr

  • MDL number MFCD09842810
  • Cas Number : 114414-17-4
  • Molecular Weight : 269.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2471
Availability details for
Quantity
  • Availability
  • Add to Cart